Methyl-3-(4-nitrophenyl)propionoate ≥97%

Supplier: Matrix Scientific

083314-1G
102839-432EA 863.08 USD
102839-432
Methyl-3-(4-nitrophenyl)propionoate ≥97%
Methyl-3-(4-nitrophenyl)propionoate

Hazard: Irritant

Formula: C₁₀H₁₁NO₄
MDL Number: MFCD00126837
CAS Number: 54405-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR