L-Phenylalanine ethyl ester hydrochloride ≥97%
Supplier: Matrix Scientific
Synonyms:
H-Phe-OEt·HCl
, Ethyl L-phenylalaninate hydrochloride
083287-1G
083287-10G
102839-378EA
116.92
USD
102839-378
102839-376
L-Phenylalanine ethyl ester hydrochloride ≥97%
L-Phenylalanine ethyl ester hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₆ClNO₂ MW: 229.704 g/mol |
MDL Number:
MFCD00012507 CAS Number: 3182-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...