Methyl-2-bromo-4,5-difluorobenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4,5-difluorobenzoic acid methyl esterMethyl 2-bromo-4,5-difluorobenzoate

083276-1G
102839-346EA 636.92 USD
102839-346
Methyl-2-bromo-4,5-difluorobenzoate ≥97%
Methyl-2-bromo-4,5-difluorobenzoate

Hazard: Irritant

Formula: C₈H₅BrF₂O₂
MW: 251.03 g/mol
MDL Number: MFCD08458104
CAS Number: 878207-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR