4-Methylnicotinoyl chloride ≥95%

Supplier: Matrix Scientific

083135-1G
102839-008EA 723.08 USD
102839-008
4-Methylnicotinoyl chloride ≥95%
4-Methylnicotinoyl chloride

Hazard: Irritant

Formula: C₇H₆ClNO
MDL Number: MFCD08751335
CAS Number: 155136-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR