2,4-Dimethoxybenzoyl chloride ≥97%

Supplier: Matrix Scientific

083099-10G 083099-1G
102838-910EA 101.54 USD
102838-910 102838-912
2,4-Dimethoxybenzoyl chloride ≥97%
2,4-Dimethoxybenzoyl chloride

Hazard: Irritant

Formula: C₉H₉ClO₃
MW: 200.62 g/mol
MDL Number: MFCD00017527
CAS Number: 39828-35-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR