3,4-Dimethylbenzoyl chloride ≥97%

Supplier: Matrix Scientific

083086-1G 083086-10G
102838-882EA 241.54 USD
102838-882 102838-884
3,4-Dimethylbenzoyl chloride ≥97%
3,4-Dimethylbenzoyl chloride

Hazard: Irritant

Formula: C₉H₉ClO
MW: 168.62 g/mol
CAS Number: 21900-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR