Pyridine-2,3-dicarbonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2,3-Dicyanopyridine

085076-1G
102844-180EA 383.08 USD
102844-180
Pyridine-2,3-dicarbonitrile ≥97%
Pyridine-2,3-dicarbonitrile

Hazard: Irritant

Formula: C₇H₃N₃
MW: 129.12 g/mol
MDL Number: MFCD00191682
CAS Number: 17132-78-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR