Ethionamide ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Ethylpyridine-4-carbothioamide2-Ethylthioisonicotinamide

085028-1G 085028-10G 085028-50G
102844-070EA 101.54 USD
102844-070 102844-072 102844-074
Ethionamide ≥97%
Ethionamide

Purity: 97%
CAS: 536-33-4
MDL Number: MFCD00057361
Molecular Formula: C8H10N2S
Molecular Weight: 166,25
Hazard: Irritant

Formula: C₈H₁₀N₂S
MW: 166.24 g/mol
MDL Number: MFCD00057361
CAS Number: 536-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR