1-(3-(Trifluoromethyl)-2-pyridinyl)piperazine hydrochloride ≥97%
Supplier: Matrix Scientific
Synonyms:
2-(Piperazin-1-yl)-3-(trifluoromethyl)pyridine hydrochloride
085018-1G
102844-046EA
200
USD
102844-046
1-(3-(Trifluoromethyl)-2-pyridinyl)piperazine hydrochloride ≥97%
1-(3-(Trifluoromethyl)-2-pyridinyl)piperazine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₂F₃N₃.HCl MW: 267.68 g/mol |
MDL Number:
MFCD00114705 CAS Number: 87394-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...