2,4-Dichloro-6-methylbenzenesulfonamide ≥85%

Supplier: Matrix Scientific

085004-1G
102843-998EA 286.35 USD
102843-998
2,4-Dichloro-6-methylbenzenesulfonamide ≥85%
2,4-Dichloro-6-methylbenzenesulphonamide

Hazard: Irritant

Formula: C₇H₇Cl₂NO₂S
CAS Number: 568577-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR