4-Fluorothiophenol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Fluorobenzenethiol4-Fluorobenzene-1-thiol1-Fluoro-4-mercaptobenzenep-Fluorothiophenol

084981-50G 084981-10G
102843-908EA 64.62 USD
102843-908 102843-910
4-Fluorothiophenol ≥97%
4-Fluorothiophenol

Hazard: Flammable/Toxic/Stored under Argon/Air Sensitive/Refrigerate

Formula: C₆H₅FS
MW: 128.17 g/mol
Boiling Pt: 170…2 °C
Density: 1.203
Flash Pt: 57 °C
MDL Number: MFCD00004846
CAS Number: 371-42-6
UN: 3071
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR