3,4-Dichlorobenzonitrile ≥95%
Supplier: Matrix Scientific
084952-1G
084952-10G
084952-50G
102843-820EA
1090.77
USD
102843-820
102843-818
102843-816
3,4-Dichlorobenzonitrile ≥95%
3,4-Dichlorobenzonitrile
Hazard: Irritant
Formula:
C₇H₃Cl₂N MW: 172.01 g/mol Melting Pt: 69…74 °C |
MDL Number:
MFCD00016379 CAS Number: 6574-99-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...