3-(1-Pyrrolyl)benzonitrile ≥97%

Supplier: Matrix Scientific

084891-1G
102843-660EA 476.1 USD
102843-660
3-(1-Pyrrolyl)benzonitrile ≥97%
3-(1-Pyrrolyl)benzonitrile

Hazard: Irritant

Formula: C₁₁H₈N₂
MDL Number: MFCD00052877
CAS Number: 175134-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR