3-Chloro-1-(2,5-dimethoxyphenyl)propan-1-one ≥97%

Supplier: Matrix Scientific

084886-1G
102843-650EA 666.15 USD
102843-650
3-Chloro-1-(2,5-dimethoxyphenyl)propan-1-one ≥97%
3-Chloro-1-(2,5-dimethoxyphenyl)propan-1-one

Hazard: Irritant

Formula: C₁₁H₁₃ClO₃
MDL Number: MFCD00126487
CAS Number: 50786-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR