(3-Bromophenyl)hydrazine ≥97%

Supplier: Matrix Scientific

084870-1G
102843-610EA 193.85 USD
102843-610
(3-Bromophenyl)hydrazine ≥97%
(3-Bromophenyl)hydrazine

Hazard: Irritant

Formula: C₆H₇BrN₂
MDL Number: MFCD02656654
CAS Number: 40887-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR