6-Bromo-m-toluonitrile ≥97%

Supplier: Matrix Scientific

084869-1G 084869-10G
102843-606EA 101.54 USD
102843-606 102843-608
6-Bromo-m-toluonitrile ≥97%
6-Bromo-m-toluonitrile

Hazard: Irritant

Formula: C₈H₆BrN
MW: 196.05 g/mol
MDL Number: MFCD11040281
CAS Number: 42872-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR