3,4-Diaminobenzimidamide ≥97%
Supplier: Matrix Scientific
084863-1G
102843-592EA
863.08
USD
102843-592
3,4-Diaminobenzimidamide ≥97%
3,4-Diaminobenzimidamide
Hazard: Irritant
Formula:
C₇H₁₀N₄ MW: 150.1811 g/mol |
MDL Number:
MFCD08751343 CAS Number: 68827-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...