Chloromethyl phenyl sulfone ≥97%

Supplier: Matrix Scientific

Synonyms: ((Chloromethyl)sulphonyl)benzenePhenyl Chloromethyl sulfone1-(Chloromethylsulfonyl)benzenePhenyl Chloromethyl Sulphone((Chloromethyl)sulfonyl)benzene1-(Chloromethylsulphonyl)benzene

084862-10G 084862-1G
102843-588EA 184.62 USD
102843-588 102843-590
Chloromethyl phenyl sulfone ≥97%
Chloromethyl phenyl sulfone

Purity: 97%
CAS: 7205-98-3
MDL Number: MFCD00007551
Molecular Formula: C7H7ClO2S
Molecular Weight: 190,65
Hazard: Irritant

Formula: C₇H₇ClO₂S
MW: 190.64 g/mol
MDL Number: MFCD00007551
CAS Number: 7205-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR