4'-Morpholinoacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-[4-(Morpholin-4-yl)phenyl]ethan-1-one

084859-1G 084859-10G
102843-576EA 101.54 USD
102843-576 102843-578
4'-Morpholinoacetophenone ≥97%
4'-Morpholinoacetophenone

Hazard: Irritant

Formula: C₁₂H₁₅NO₂
MW: 205.2530 g/mol
MDL Number: MFCD00006168
CAS Number: 39910-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR