2-Propylphenol ≥97%
Supplier: Matrix Scientific
Synonyms:
1-Hydroxy-2-propylbenzene
084857-1G
102843-574EA
86.15
USD
102843-574
2-Propylphenol ≥97%
2-Propylphenol
Hazard: Irritant
Formula:
C₉H₁₂O MW: 136.19 g/mol |
MDL Number:
MFCD00002251 CAS Number: 644-35-9 UN: 3145 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...