1-[2-Nitro-5-(trifluoromethyl)phenyl]-1H-pyrrole ≥97%

Supplier: Matrix Scientific

084855-1G
102843-570EA 410.77 USD
102843-570
1-[2-Nitro-5-(trifluoromethyl)phenyl]-1H-pyrrole ≥97%
1-[2-Nitro-5-(trifluoromethyl)phenyl]-1H-pyrrole

Hazard: Irritant

Formula: C₁₁H₇F₃N₂O₂
MDL Number: MFCD22566369
CAS Number: 1795460-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR