3',5'-Dichloro-2,2,2-trifluoroacetophenone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(3,5-Dichlorophenyl)-2,2,2-trifluoroethanone
084834-1G
102843-510EA
44.62
USD
102843-510
3',5'-Dichloro-2,2,2-trifluoroacetophenone ≥95%
3',5'-Dichloro-2,2,2-trifluoroacetophenone
Hazard: Irritant
Formula:
C₈H₃Cl₂F₃O MW: 243.0100 g/mol |
MDL Number:
MFCD01319996 CAS Number: 130336-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...