Pentachlorobenzonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2,3,4,5,6-Pentachlorobenzonitrile

084771-10G 084771-1G
102843-334EA 1163.08 USD
102843-334 102843-332
Pentachlorobenzonitrile ≥97%
Pentachlorobenzonitrile

Hazard: Irritant

Formula: C₇Cl₅N
MW: 275.35 g/mol
MDL Number: MFCD00173666
CAS Number: 20925-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR