2'-Fluoro-5'-nitroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(2-Fluoro-5-nitrophenyl)ethanone

084766-1G
102843-316EA 44.85 USD
102843-316
2'-Fluoro-5'-nitroacetophenone ≥97%
2'-Fluoro-5'-nitroacetophenone

Hazard: Irritant

Formula: C₈H₆FNO₃
MW: 183.1400 g/mol
MDL Number: MFCD00277867
CAS Number: 79110-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR