2-Aminobenzenesulfonic acid ≥97%

Supplier: Matrix Scientific

Synonyms: Aniline-2-sulphonic acidOrthanilic acid

084756-50G
102843-286EA 21.54 USD
102843-286
2-Aminobenzenesulfonic acid ≥97%
2-Aminobenzenesulfonic acid

Hazard: Irritant

Formula: C₆H₇NO₃S
MDL Number: MFCD00007705
CAS Number: 88-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR