1-Chloro-4-[(2-chloroethyl)sulfonyl]benzene ≥97%

Supplier: Matrix Scientific

084755-1G
102843-284EA 101.54 USD
102843-284
1-Chloro-4-[(2-chloroethyl)sulfonyl]benzene ≥97%
1-Chloro-4-[(2-chloroethyl)sulphonyl]benzene

Hazard: Irritant

Formula: C₈H₈Cl₂O₂S
MDL Number: MFCD00018966
CAS Number: 16191-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR