(4-Methoxyphenoxy)thioacetamide ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(4-Methoxyphenoxy)ethanethioamide

084721-1G
102843-192EA 241.54 USD
102843-192
(4-Methoxyphenoxy)thioacetamide ≥97%
(4-Methoxyphenoxy)thioacetamide

Hazard: Irritant

Formula: C₉H₁₁NO₂S
MDL Number: MFCD00052482
CAS Number: 35370-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR