4-Benzyloxybenzotrifluoride ≥97%

Supplier: Matrix Scientific

084701-1G
102843-146EA 264.5 USD
102843-146
4-Benzyloxybenzotrifluoride ≥97%
4-Benzyloxybenzotrifluoride

Hazard: Irritant

Formula: C₁₄H₁₁F₃O
MW: 252.23
MDL Number: MFCD00275905
CAS Number: 70097-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR