3-Bromo-2-nitroanisole ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-3-methoxy-2-nitrobenzene

084686-1G
102843-092EA 467.69 USD
102843-092
3-Bromo-2-nitroanisole ≥97%
3-Bromo-2-nitroanisole

Hazard: Irritant

Formula: C₇H₆BrNO₃
MW: 232.0314 g/mol
MDL Number: MFCD11101044
CAS Number: 500298-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR