2'-Fluoroacetanilide ≥97%

Supplier: Matrix Scientific

084669-50G
102843-046EA 232.31 USD
102843-046
2'-Fluoroacetanilide ≥97%
2'-Fluoroacetanilide

Hazard: Irritant

Formula: C₈H₈FNO
MW: 153.16 g/mol
MDL Number: MFCD00000315
CAS Number: 399-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR