4-Amino-N-(2,4-difluorophenyl)benzenesulfonamide ≥97%

Supplier: Matrix Scientific

084660-1G
102843-024EA 666.15 USD
102843-024
4-Amino-N-(2,4-difluorophenyl)benzenesulfonamide ≥97%
4-Amino-N-(2,4-difluorophenyl)benzenesulphonamide

Hazard: Irritant

Formula: C₁₂H₁₀F₂N₂O₂S
MDL Number: MFCD01935782
CAS Number: 1717-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR