5-Nitro-2-(1-pyrrolyl)benzonitrile ≥97%

Supplier: Matrix Scientific

084631-1G
102842-938EA 392.15 USD
102842-938
5-Nitro-2-(1-pyrrolyl)benzonitrile ≥97%
5-Nitro-2-(1-pyrrolyl)benzonitrile

Hazard: Irritant

Formula: C₁₁H₇N₃O₂
MDL Number: MFCD00052251
CAS Number: 106981-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR