1-(2,4-Dimethylphenyl)-2-thiourea ≥98%

Supplier: Matrix Scientific

Synonyms: 2,4-DimethylphenylthioureaN-(2,4-Dimethylphenyl)thiourea1-(2,4-Dimethylphenyl)thiourea1-(2,4-Xylyl)thiourea

084572-10G
102842-790EA 246.15 USD
102842-790
1-(2,4-Dimethylphenyl)-2-thiourea ≥98%
1-(2,4-Dimethylphenyl)-2-thiourea

Hazard: Irritant

Formula: (C₆H₃(CH₃)₂)NC(S)NH₂
MW: 180.27 g/mol
MDL Number: MFCD00041164
CAS Number: 16738-20-8
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR