2-(1-Pyrrolidinyl)phenol ≥97%

Supplier: Matrix Scientific

084548-1G 084548-10G
102842-718EA 676.2 USD
102842-718 102842-716
2-(1-Pyrrolidinyl)phenol ≥97%
2-(1-Pyrrolidinyl)phenol

Hazard: Irritant

Formula: C₁₀H₁₃NO
MDL Number: MFCD00051659
CAS Number: 4787-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR