6-Chloro-4-nitro-o-cresol ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-methyl-4-nitrophenol

084546-1G
102842-712EA 1287.69 USD
102842-712
6-Chloro-4-nitro-o-cresol ≥97%
6-Chloro-4-nitro-o-cresol

Hazard: Irritant

Formula: C₇H₆ClNO₃
MDL Number: MFCD00047740
CAS Number: 4102-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR