2-Cyanobenzenesulfonamide ≥97%

Supplier: Matrix Scientific

084535-1G 084535-10G
102842-678EA 104.62 USD
102842-678 102842-680
2-Cyanobenzenesulfonamide ≥97%
2-Cyanobenzenesulphonamide

Hazard: Irritant

Formula: C₇H₆N₂O₂S
MW: 182.2 g/mol
Melting Pt: 160…164 °C
MDL Number: MFCD03094593
CAS Number: 73542-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR