5'-Acetamido-2'-hydroxyacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: N-(3-Acetyl-4-hydroxyphenyl)acetamide

084533-1G
102842-672EA 144.62 USD
102842-672
5'-Acetamido-2'-hydroxyacetophenone ≥97%
5'-Acetamido-2'-hydroxyacetophenone

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MW: 193.2000 g/mol
MDL Number: MFCD00100490
CAS Number: 7298-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR