4-(Pentafluorothio)benzonitrile ≥97%

Supplier: Matrix Scientific

084518-1G
102842-634EA 124.2 USD
102842-634
4-(Pentafluorothio)benzonitrile ≥97%
4-(Pentafluorothio)benzonitrile

Hazard: Irritant

Formula: C₇H₄F₅NS
MDL Number: MFCD03788521
CAS Number: 401892-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR