2,6-Dichloro-4-nitrotoluene ≥97%
Supplier: Matrix Scientific
Synonyms:
1,3-Dichloro-2-methyl-5-nitrobenzene
084490-10G
084490-1G
102842-552EA
255.38
USD
102842-552
102842-554
2,6-Dichloro-4-nitrotoluene ≥97%
2,6-Dichloro-4-nitrotoluene
Hazard: Irritant
Formula:
C₇H₅Cl₂NO₂ MW: 206.03 g/mol |
MDL Number:
MFCD00174267 CAS Number: 7149-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...