4-Nitro-o-toluonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Nitro-o-tolunitrile2-Methyl-4-nitrobenzonitrile

084482-1G
102842-532EA 255.38 USD
102842-532
4-Nitro-o-toluonitrile ≥97%
4-Nitro-o-toluonitrile

Hazard: Irritant

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD03095378
CAS Number: 89001-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR