3-Methyl-2-nitroanisole ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Methoxy-3-methyl-2-nitrobenzeneMethyl 2-nitro-m-tolyl ether3-Methoxy-2-nitrotoluene

084459-50G 084459-10G
102842-458EA 129.23 USD
102842-458 102842-460
3-Methyl-2-nitroanisole ≥97%
3-Methyl-2-nitroanisole

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00007179
CAS Number: 5345-42-6
UN: 2446
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR