2,1,3-Benzothiadiazole-5-sulfonyl chloride ≥85%

Supplier: Matrix Scientific

084451-1G
102842-442EA 527.69 USD
102842-442
2,1,3-Benzothiadiazole-5-sulfonyl chloride ≥85%
2,1,3-Benzothiadiazole-5-sulfonyl chloride

Hazard: Irritant

Formula: C₆H₃ClN₂O₂S₂
MDL Number: MFCD02681993
CAS Number: 337508-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR