3-Benzo[b]thiophenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: Benzothiophene-3-sulfonyl chloride1-Benzothiophene-3-sulfonyl Chloride1-Benzothiophene-3-sulphonyl ChlorideBenzothiophene-3-sulphonyl chloride

084446-1G
102842-432EA 353.85 USD
102842-432
3-Benzo[b]thiophenesulfonyl chloride ≥97%
3-Benzo[b]thiophenesulphonyl chloride

Hazard: Corrosive

Formula: C₈H₅ClO₂S₂
MW: 232.71 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD02681897
CAS Number: 18494-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR