3-(Chloromethyl)-5-phenyl-1,2,4-oxadiazole ≥97%

Supplier: Matrix Scientific

084364-1G
102842-206EA 152.31 USD
102842-206
3-(Chloromethyl)-5-phenyl-1,2,4-oxadiazole ≥97%
3-(Chloromethyl)-5-phenyl-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₉H₇ClN₂O
MW: 194.6177 g/mol
MDL Number: MFCD00084968
CAS Number: 1201-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR