(±)-Tetrahydrofurfuryl bromide ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Bromomethyl)tetrahydrofuran

084351-10G 084351-50G 084351-1G
102842-178EA 384.38 USD
102842-178 102842-174 102842-176
(±)-Tetrahydrofurfuryl bromide ≥97%
(±)-Tetrahydrofurfuryl bromide

Hazard: Irritant

Formula: C₅H₉BrO
MW: 165.04
MDL Number: MFCD00016894
CAS Number: 1192-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR