MATR084342

084342-50G
102842-150EA 49.69 USD
102842-150
MATR084342
Purity: 97%
CAS: 459-46-1
MDL Number: MFCD00000359
Molecular Formula: C7H6BrF
Molecular Weight: 189,03
Boiling Point: 85°C/15mmHg
Hazard: Corrosive/Lachrymator
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR