4-Phenoxybenzyl bromide ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(Bromomethyl)-4-phenoxybenzene

084328-1G
102842-112EA 553.85 USD
102842-112
4-Phenoxybenzyl bromide ≥97%
4-Phenoxybenzyl bromide

Hazard: Irritant

Formula: C₁₃H₁₁BrO
MW: 263.13 g/mol
MDL Number: MFCD04038693
CAS Number: 36881-42-2
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR