α-Bromo-m-xylene ≥98%
Supplier: Matrix Scientific
Synonyms:
3-Methylbenzyl bromide
084321-10G
084321-50G
084321-1G
102842-090EA
214.39
USD
102842-090
102842-086
102842-088
α-Bromo-m-xylene ≥98%
α-Bromo-m-xylene
Hazard: Irritant
Formula:
C₈H₉Br MW: 185.06 g/mol |
MDL Number:
MFCD00000177 CAS Number: 620-13-3 UN: 1701 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...