2,6-Dimethoxyphenyl isocyanate ≥95%

084306-1G
102842-038EA 1055.67 USD
102842-038
2,6-Dimethoxyphenyl isocyanate ≥95%
2,6-Dimethoxyphenyl isocyanate

Hazard: Irritant

Formula: C₉H₉NO₃
MW: 179.18 g/mol
CAS Number: 50528-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR