4-Fluoro-3-methoxybenzyl alcohol ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Fluoro-3-methoxyphenyl)methanol

084173-1G
102841-714EA 121.54 USD
102841-714
4-Fluoro-3-methoxybenzyl alcohol ≥97%
4-Fluoro-3-methoxybenzyl alcohol

Hazard: Irritant

Formula: C₈H₉FO₂
MW: 156.16 g/mol
MDL Number: MFCD00272133
CAS Number: 128495-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR